Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sklar Blue™ Graves Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5024 91-5018 91-5066 91-5028 91-5012 91-5062 91-5014 91-5060 91-5015 91-5068 91-5020 91-5010 91-5064 91-5080 91-5022
10194-316EA 400.32 USD InStock
10194-316 10193-372 10184-240 10184-394 10194-138 10184-246 10184-400 10194-030 10184-242 10184-396 10184-398 10184-244 10184-442 10193-322 10184-238
Sklar Blue™ Graves Vaginal Speculum, OR Grade, Sklar
Specula
Sklar Blue™ Graves Vaginal Speculum is used for electrocsurgical gynecological procedures, such as LEEP.
  • Feature high quality stainless steel
  • With an insulated coating
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. It has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient. It is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation.
This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue instruments are fully autoclavable and may also be cleaned ultrasonically.
Certifications: All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty