Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sklar Blue™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5122 91-5132
10184-456EA 1505.26 USD InStock
10184-456 10184-452
Sklar Blue™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Specula
These are single end vaginal speculum commonly used in electrosurgical OB/GYN procedures.
  • Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating
  • Safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue
  • Fully autoclavable and may also be cleaned ultrasonically
Available with various blades styles and weight choices in order to accommodate the needs of the patient and the procedure. The weight (21/2 lbs.) holds the speculum in place and provides an open view for the surgeon to perform procedures.
Comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty