Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sklar Blue™ Steiner-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5142 91-5140
10184-460EA 1391.54 USD InStock
10184-460 10184-458
Sklar Blue™ Steiner-Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Blue™ Steiner-Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in OB/GYN procedures such as vaginal hysterectomy or dilation and curettage.
  • It is weighted with a medium, 21/2 lbs.
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
Certifications: Comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty