Would you like to log in again?
Sklar Blue™ Cusco Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5152 91-5150 91-5154
10184-466EA 469.27 USD InStock
10184-466 10184-462 10184-464
Sklar Blue™ Cusco Vaginal Speculum, OR Grade, Sklar
Specula
The Cusco vaginal speculum with lateral screw is a self-retaining speculum used for examination of, or procedures within, the vaginal canal.
  • The blades of the Cusco speculum are opened, closed, and adjusted with the attached screw
  • Electrosurgical instrument feature high quality stainless steel with an insulated coating
This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty