Would you like to log in again?
Sklar Blue™ Emmett Hook, OR Grade, Sklar
Supplier: Sklar
91-5253 91-5251 91-5250 91-5254 91-5252
10184-510EA 260.74 USD InStock
10184-510 10184-506 10184-508 10184-502 10184-504
Sklar Blue™ Emmett Hook, OR Grade, Sklar
Dissection Needles
The Sklar Blue™ Emmett Hook, which may also be known as an Emmett Iris Hook, is most commonly used in electrosurgical OB/GYN procedures involving the uterus.
  • The hook comes in a variety of styles
  • Reusable
  • Latex-free
Sklar Blue™ instruments feature high quality stainless steel with an insulated coating that prevents radio frequency currents from being transmitted through the instrument and thus reducing the burning of non-targeted tissue. Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty