Would you like to log in again?
Sklar Blue™ Tuttle Forceps, OR Grade, Sklar
Supplier: Sklar
91-5300
10184-518EA 392.25 USD InStock
10184-518
Sklar Blue™ Tuttle Forceps, OR Grade, Sklar
Forceps
Sklar Blue™ Tuttle Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
These forceps have long shafts which make them useful for working in areas such as the intestines. These forceps may also be known as Singley forceps. Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument, which reduces the burning of non-targeted tissue.
Certifications: Sklar Blue™ instruments are autoclavable, and may also be cleaned ultrasonically. All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty