Would you like to log in again?
Sklar Blue™ Laufe Uterine Polyp Forceps, OR Grade, Sklar
Supplier: Sklar
91-5408
10184-530EA 456.68 USD InStock
10184-530
Sklar Blue™ Laufe Uterine Polyp Forceps, OR Grade, Sklar
Forceps
Sklar Blue™ Laufe Uterine Polyp Forceps are non-conductive Nylon coated and industry standard.
Certifications: All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty