Would you like to log in again?
Merit™ Operating Scissors, Physician Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Merit™
97-292 97-299 97-269 97-266 97-275 97-281 97-263 97-272 97-295 97-284 97-270 97-278
10193-704EA 10.75 USD InStock
10193-704 10184-600 10193-682 10194-480 10193-642 10184-606 10193-872 10184-608 10184-602 10184-404 10184-406 10184-604
Merit™ Operating Scissors, Physician Grade, Sklar
Scissors
Merit™ operating scissors, also known as surgical scissors are a versatile surgical instrument
Merit™ instruments are office-grade line of products used for routine and in-office minor procedures.
These high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
They are used for general, multipurpose cutting and dissecting. Sharp operating scissors cut tougher tissue and are frequently used for dissecting, whereas blunt operating scissors are more often used for cutting materials such as sutures, tubing, or gauze.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty