Would you like to log in again?
Surgi-OR™ Semken Dressing Forceps, Mid-Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-478
10184-612EA 34.53 USD InStock
10184-612
Surgi-OR™ Semken Dressing Forceps, Mid-Grade, Sklar
Forceps
Surgi-OR™ Semken Dressing Forceps are most commonly used for application and changing of dressings.
These forceps feature narrow tips that may be beneficial for neurosurgery or other procedures performed in constricted areas. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty