Would you like to log in again?
Curette Excavators, Rounded, OR Grade, Sklar
Supplier: Sklar
97-0507 97-0509 97-0503 97-0505
10184-696EA 62.42 USD InStock
10184-696 10184-692 10184-694 10184-690
Curette Excavators, Rounded, OR Grade, Sklar
Curettes
Sklar's® Curette Excavators are commonly used for scraping tissue.
These are available in single and double-ended styles with or without fenestrated cupped tips in various sizes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty