Would you like to log in again?
Moleskin Scissors, OR Grade, Sklar
Supplier: Sklar
97-1351 97-1350
10184-756EA 251.67 USD InStock
10184-756 10184-758
Moleskin Scissors, OR Grade, Sklar
Scissors
Moleskin Scissors are non-ratcheted, finger ring scissors often used for cutting moleskin, a cotton fabric frequently used in the prevention and treatment of blisters, corns, or calluses
These scissors have one sharp tip and one blunt tip. The edge of the blade with the pointed tip is serrated, while the edge of the blade with the blunt tip is smooth. They are available straight or curved.
The fabric can be cut to fit the size of the area in which it is needed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty