Would you like to log in again?
Tungsten Carbide Allis Forceps, OR Grade, Sklar
Supplier: Sklar
97-1560 97-1555
10184-774EA 503.86 USD InStock
10184-774 10184-776
Tungsten Carbide Allis Forceps, OR Grade, Sklar
Forceps
Tungsten Carbide Atraumatic Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
TC instruments are identified by gold handles.
They are used with tonsil, vaginal, breast, and thyroid tissue. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty