Would you like to log in again?
Babcock Tungsten Carbide Forceps, OR Grade, Sklar
Supplier: Sklar
97-1565 97-1570
10184-780EA 581.35 USD InStock
10184-780 10184-778
Babcock Tungsten Carbide Forceps, OR Grade, Sklar
Forceps
Babcock Tungsten Carbide Forceps are finger ring, ratcheted, non-perforating forceps used to grasp delicate tissue
Babcock Forceps are similar to Allis forceps; however, may be considered less traumatic due to their wider, rounded grasping surface. The jaws are circumferential, and the tips are triangular and fenestrated. TC Babcock forceps have cross serrations. They are particularly useful for grasping tube-shaped structures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
They are used with intestinal and laparotomy procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty