Would you like to log in again?
Merit™ Rankin-Kelly Hemostatic Forceps, Physician Grade, Sklar
Supplier: Sklar
Merit™
97-469 97-465
10184-810EA 30.2 USD InStock
10184-810 10184-812
Merit™ Rankin-Kelly Hemostatic Forceps, Physician Grade, Sklar
Forceps
Merit™ Rankin-Kelly Forceps are used for clamping large blood vessels or manipulating heavy tissue
They are available curved or straight with a ratcheted finger ring handle. Rankin and Kelly forceps are almost identical. The jaws of both are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Rankin forceps are longer and lighter than Kelly forceps.
Merit™ instruments are mid-grade products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
They may also be used for soft tissue dissection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty