Would you like to log in again?
Merit™ Rochester-Pean Forceps, Physician Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Merit™
97-492 97-483 97-487 97-481 97-496 97-470 97-471 97-494 97-473 97-485 97-498 97-477
10184-830EA 33.96 USD InStock
10184-830 10184-820 10184-818 10184-814 10184-826 10184-816 10184-828 10184-832 95042-906 10184-822 10184-834 10184-824
Merit™ Rochester-Pean Forceps, Physician Grade, Sklar
Forceps
Merit™ Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight.
Merit™ instruments are mid-grade products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty