Would you like to log in again?
Merit™ Adson-Brown Tissue Forceps, Physician Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Merit™
97-603 97-778
10184-872EA 42.37 USD InStock
10184-872 10184-908
Merit™ Adson-Brown Tissue Forceps, Physician Grade, Sklar
Forceps
Merit™ Adson-Brown Tissue Forceps are for holding and manipulating delicate tissues.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, typically 7 x 7 or 9 x 9. They are used to grasp delicate tissue, most commonly in plastic surgery. Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures.
High quality instruments crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices, that require quality, durable instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty