Would you like to log in again?
Corning® CellBIND® 384-Well, Clear Treated Microplate, Corning
Supplier: Corning
Corning®
4579
10185-092CS 1042.78 USD InStock
10185-092
Corning® CellBIND® 384-Well, Clear Treated Microplate, Corning
Microplates Cell Culture Plates
The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium
This enhancement results in higher cell yields.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty