Would you like to log in again?
Merit™ Littauer Cutting Forceps, Physician Grade, Sklar
Supplier: Sklar
Merit™
98-476 98-477 98-478 98-475
10185-132EA 115.4 USD InStock
10185-132 10185-136 10185-134 10185-138
Merit™ Littauer Cutting Forceps, Physician Grade, Sklar
Bone Cutters and Rongeurs
Merit™ Littauer Cutting forceps are office-grade, non-ratcheted, plier handle instrument
The short, sharp blades are able to produce enough force to cut through thick nail and small bone, but delicate enough not to cause trauma to surrounding tissue or organs. This is helpful with nail debridement, which should be completed in short, small sections to avoid damage to the hyponychium.
As nail nippers they are used for nail debridement. They may also be used on delicate bones found in the facial and nasal region, or small animal (mice) veterinary procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty