Would you like to log in again?
Micro Scissors, Sklar
Supplier: Sklar
98-7814 98-7837 98-7813 98-7892
75996-308EA 875.89 USD InStock
75996-308 75996-306 75996-324 10185-190
Micro Scissors, Sklar
Scissors
Sklar's® micro scissors are used for the dissection of tissues, blood vessels and nerves.
  • Reusable
  • Nonsterile
Available with a flat or round handle. The straight scissors may be used to cut sutures and trim nerve endings. Curved will dissect vessels and nerves. In addition to the tips, the sides may be used for dissection with the tips closed, similar to using forceps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty