Would you like to log in again?
TC Hegar Vascular Needle Holder, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20-4178 20-4175
10185-274EA 347.33 USD InStock
10185-274 10195-176
TC Hegar Vascular Needle Holder, OR Grade, Sklar
Forceps
Sklar's® TC Hegar finger ring needle holders are used in cardiovascular surgeries for suturing.
Tungsten carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments
TC Hegar and Censitrac Ready products are industry standard and constructed of stainless steel.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty