Would you like to log in again?
Half Circle Fistula Needle, OR Grade, Sklar
Supplier: Sklar
93-1710-6 93-1710-1 93-1710-00 93-1710-2 93-1710-8
10185-642PK 29.65 USD InStock
10185-642 10185-640 10185-646 10185-644 10185-648
Half Circle Fistula Needle, OR Grade, Sklar
Syringe Needles
Sklar's Half Circle Stainless Steel Fistula Needle is used in veterinary suturing procedures.
The half circle is useful in confined surgical sites, although it requires the user to move his wrist more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty