Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ferguson Suture Needles, OR Grade, Sklar
Supplier: Sklar
93-1716-14 93-1716-16 93-1716-10 93-1716-12 93-1716-4 93-1716-2 93-1716-8 93-1716-6
10185-682PK 35.35 USD InStock
10185-682 10185-672 10185-676 10185-674 10185-404 10185-688 10185-402 10185-678
Ferguson Suture Needles, OR Grade, Sklar
Dissection Needles
Ferguson Suture Needle is used for suturing organs and vessels in veterinary procedures.
The half circle curvature of this needle makes it useful for suturing in difficult to reach areas, but requires placement with a needle holder or other holding instrument. Ferguson needle has a taper point and is available in several sizes to accomodate a variety of tissue as well as the type of suture material being used.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty