Would you like to log in again?
Post Mortem Needles, OR Grade, Sklar
Supplier: Sklar
93-1731-2 93-1731-1 93-1731-4 93-1731-3
10185-694PK 79.12 USD InStock
10185-694 10185-698 10185-696 10185-700
Post Mortem Needles, OR Grade, Sklar
Dissection Needles
Half Circle Post Mortem Needles are used for suturing in autopsy, necropsy, and other forensic procedures in veterinary practice.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty