Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Regular Surgeon Needles, Spring Eye, OR Grade, Sklar
Supplier: Sklar
93-1760-4 93-1760-6 93-1760-10 93-1760-1 93-1760-14 93-1760-2 93-1760-8 93-1760-16 93-1760-12
10185-770PK 29.65 USD InStock
10185-770 10185-782 10185-780 10185-774 10185-772 10185-778 10185-768 10185-776 10185-766
Regular Surgeon Needles, Spring Eye, OR Grade, Sklar
Dissection Needles
Regular Surgeons Needles are used for common suturing in veterinary procedures.
This product is a 3/8 circle needle with a cutting edge and a spring eye.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty