Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ready-to-use Cell Proliferation Colorimetric Reagent, WST-1, Biovision

Supplier: BioVision
AB155902-2500TEST
10186-612EA 1038.18 USD InStock
10186-612
Ready-to-use Cell Proliferation Colorimetric Reagent, WST-1, Biovision
Assays Cellular Assays
Sensitive, Non-radioactive Assay

Detection method- Absorbance 440 nm
Species reactivity- Mammalian
Applications- Measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients, etc. Analysis of cytotoxic and cytostatic compounds such as anticancer drugs, toxic agents and other pharmaceuticals. Assessment of physiological mediators and antibodies that inhibit cell growth.

The ready-to-use cell proliferation reagent, WST-1 provides a simple and accurate method to measure cell proliferation, which is based on the cleavage of the tetrazolium salt WST-1 to formazan by cellular mitochondrial dehydrogenases. Expansion in the number of viable cells results in an increase in the activity of the mitochondrial dehydrogenases, which in turn leads to increase in the amount of formazan dye formed. The formazan dye produced by viable cells can be quantified by measuring the absorbance at 440 nm. This new method is non-radioactive, rapid and more sensitive than MTT, XTT, or MTS-based assays. The entire assay can be performed in the same microtiter plate and does not require extra steps like washing, harvesting and cell solubilization.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty