Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Akt Activity Assay Kit, KinaseSTAR, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AB65786-40
10186-710EA 759.54 USD InStock
10186-710
Akt Activity Assay Kit, KinaseSTAR, BioVision
Assays Cellular Assays
Specifically detects Akt1, Akt2, Akt3 activities

Detection method- Western blot analysis using anti-Phospho-GSK-3α (Ser 21) antibody at 1:1000 dilutions
Species reactivity- Mammalian
Applications- The Akt Activity Assay Kit utilizes an Akt-specific antibody to immunoprecipitate Akt from cell lysate. Activity of the Akt is then determined in a kinase reaction using recombinant GSK-3α as substrate.

Akt is a protein kinase that can be activated by insulin and various growth factors and functions in a pathway involving PI3 kinase. Recent evidence suggests that Akt functions to promote cell survival by actively inhibiting apoptosis. The Akt Activity Assay Kit utilizes an Akt-specific antibody to immunoprecipitate Akt from cell lysate. Activity of the Akt is then determined in a kinase reaction using recombinant GSK-3α as substrate. Phosphorylation of the GSK-3α can be analyzed by Western blot analysis using the phospho-GSK-3α specific antibody included in the kit. The kit specifically detects Akt1, Akt2, and Akt3 activities, other kinase activities would not be detected.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty