Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neutrophil Elastase Inhibitor Screening Kit (Fluorometric), BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AB118971-100TEST
10187-040EA 1070.89 USD InStock
10187-040
Neutrophil Elastase Inhibitor Screening Kit (Fluorometric), BioVision
Assays Cellular Assays
Detection method- Fluorescence (Ex/Em 400/505 nm)
Applications- This kit can be used for screening NE inhibitors and comes along with positive and negative control inhibitors

Neutrophil Elastase (NE) is an aggressive and cytotoxic 29 kDa serine protease stored mainly in the azurophil granules of neutrophil granulocytes. It plays a role in the degradation of a wide range of extracellular matrix proteins, including fibronectin, laminin, proteoglycans, collagens, and elastin. When the extracellular NE concentration exceeds the buffering capacity of endogenous inhibitors, it becomes implicated in the signs, symptoms and disease progression of inflammatory lung disorders via its role in the inflammatory process, mucus overproduction and lung tissue damage. In BioVision’s Neutrophil Elastase Inhibitor Screening Kit, NE hydrolyzes a specific fluorescent substrate to release the fluorescent group, which can be detected at Ex/Em = 400/505 nm. In presence of a potent Neutrophil Elastase inhibitor, the hydrolyzation of substrate will be inhibited or stopped. The kit provides a rapid, simple, sensitive, and reliable test suitable as a high throughput screening assay of Neutrophil Elastase inhibitors. For comparison of the relative efficacy of test inhibitors, a control inhibitor, SPCK (Ki = 10 µM for human leukocyte elastase) is included.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty