Would you like to log in again?
BCA Protein Assay Kit II, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB287853-2500TEST AB287853-5000TEST
10187-082EA 364.9 USD InStock
10187-082 10187-080
BCA Protein Assay Kit II, BioVision
Assays Cellular Assays
Detection method- Absorbance (562 nm)
Detergent compatible
Biovision’s BCA Protein Assay kit is useful for colorimetric detection and quantification of total protein content even in the presence of detergents. The Kit is based on the chelation of bicinchoninic acid (BCA) with the cuprous cation (Cu+1), which is generated by reduction of cupric cation (Cu+2) with the protein in an alkaline medium. The amount of purple-colored product formed during the reaction can be correlated to the amount of protein in the assay. The Cu+1-BCA chelate is a water-soluble complex and exhibits a strong absorbance at 562 nm that is linear over a wide range of protein concentrations between 25-2000 μg/ml. In general, protein concentrations are estimated with reference to a commonly used protein standard. This BCA Protein Assay Kit also includes Bovine Serum Albumin (BSA) as a protein standard for estimation of total protein content of the samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty