Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Valsartan

Synonyms: (S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid(S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid

AB141486-500MG AB141486-25MG AB141486-100MG
10188-686EA 788.83 USD InStock
10188-686 10188-688 10188-684
Valsartan
Valsartan


Formula: C₂₄H₂₉N₅O₃
MW: 435.52 g/mol
MDL Number: MFCD00865840
CAS Number: 137862-53-4
Order Now
Specification Test Results
CAS 137862-53-4
Purity ≥98% by TLC
Form White solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty