Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tween® 20 (Polysorbate) 10% in aqueous solution

Supplier: BioVision
Synonyms: Polyoxyethylene sorbitan monolaurate

AB286841-100ML
10190-930EA 184.78 USD InStock
10190-930
Tween® 20 (Polysorbate) 10% in aqueous solution
Tween® 20 (Polysorbate)


Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
MDL Number: MFCD00165986
CAS Number: 9005-64-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty