Would you like to log in again?
Ezlys™ Bacterial Protein Extraction Reagent, BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB286871-100ML AB286871-500ML
10190-950EA 458.94 USD InStock
10190-950 10190-948
Ezlys™ Bacterial Protein Extraction Reagent, BioVision
Protein Extraction Protein Extraction Reagents
A proprietary formulation prepared in a 50 mM Tris pH8.0 buffer and designed for gentle disruption of cell membrane to extract proteins. It provides a simple, rapid, low cost method for releasing expressed target protein for preparation, purification, or other applications. The EZLys™ reagent is an optically transparent, non-denaturing formulation which can be easily removed by dialysis. It is also useful for the preparation of high purity inclusion bodies for subsequent purification. This reagent does not interfere with cross linking and is suitable for use in the BCA or Bradford protein assays. Ideal pH range is pH 5 -10. The 100 ml and 500 ml reagent are sufficient for protein extraction from 25 g and 125 g of cell paste.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty