Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tween® 80 (Polysorbate) 10% in aqueous solution, MegaPure™ detergent


AB291111-500ML AB291111-50ML 2126-SET AB291111-100ML
10191-308EA 1104.73 USD InStock
10191-308 10191-306 10191-304 76289-908
Tween® 80 (Polysorbate) 10% in aqueous solution, MegaPure™ detergent
Tween® 80 (Polysorbate)
A non-ionic detergent useful for the extraction of membrane-bound proteins as well as isolation of nuclei from mammalian cell lines.

  • A non-ionic detergent


MDL Number: MFCD00082107
CAS Number: 9005-65-6
Order Now
Specification Test Results
CAS 9005-65-6
Purity Aldehydes: 200 µM; Peroxides: 200 µM
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty