Would you like to log in again?
Human Recombinant FGF18 (from E. coli)
Supplier: BioVision
AB289792-1MG
10191-898EA 6251.23 USD InStock
10191-898
Human Recombinant FGF18 (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant Human FGF-18, which plays an important role in regulating tissue growth, morphogenesis and tissue repair
The ED₅₀ as determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors is <0.5 ng/ml, corresponding to a specific activity of >2 x 10⁶ units/mg.
The FGFs plays an important role in regulating tissue growth, morphogenesis and tissue repair. FGF-18 is a newly discovered heparin binding growth factor that stimulates the proliferation and activation of cells that express FGF receptors. The human FGF-18 is a 21.2 kDa protein containing 182 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty