Would you like to log in again?
Human Recombinant IL4 (from E. coli)
Supplier: BioVision
AB285513-50UG AB285513-10UG AB285513-1MG
10191-906EA 1612.81 USD InStock
10191-906 10191-904 10191-902
Human Recombinant IL4 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant Human IL-4, which is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B cells and T cells
The ED₅₀ as determined by the dose-dependent stimulation of human TF-1 cells is <0.2 ng/ml, corresponding to a specific activity of ≥5 x 10⁶ units/mg
IL-4 is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B cells and T cells. Human IL-4 is a 15.0 kDa globular protein containing 130 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty