Would you like to log in again?
Human Recombinant IL10 (from E. coli)
Supplier: BioVision
AB285525-50UG AB285525-10UG AB285525-1MG
10191-916EA 13329.84 USD InStock
10191-916 10191-914 10190-266
Human Recombinant IL10 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant Human IL-10, which helps in inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hypersensitivity response, and stimulation of the Th2 cell response.
The ED₅₀ determined by the dose-dependent co-stimulation (with murine IL-4) of MC/9 cells is <2 ng/ml, corresponding to a specific activity of > 5 x 10⁵ units/mg.
IL-10 (Interleukin-10), originally known as Cytokine Synthesis Inhibitory Factor (CSIF), is an 18.6 kDa protein containing 161 amino acid residues. It shares over 80% sequence homology with the Epstein-Barr Virus protein BCRFI. The reported biological activities of IL-10 include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hypersensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty