Would you like to log in again?
SpectraMax® MiniMax™ 300 Imaging Cytometer, Molecular Devices
Supplier: Molecular Devices
SpectraMax®
MINIMAX300 5043091 MINIMAX300
10192-218EA 59815.88 USD InStock
CA10192-218 10192-218 76459-144
SpectraMax® MiniMax™ 300 Imaging Cytometer, Molecular Devices
Cell Counters and Flow Cytometers Flow Cytometers
Designed specifically for the SpectraMax® i3 Multi-Mode Detection Platform, the field-upgradeable SpectraMax® MiniMax™ 300 Imaging Cytometer enables quick inspection of cells and provides insight into phenotypic changes that are important for understanding cytotoxicity, cell proliferation, and protein expression.
  • Field-upgradeable
  • Quick inspection of cells
  • Enable cellular imaging
  • StainFree™ cell counting technology
Designed to enable cellular imaging capabilities, this cytometer unit now features StainFree™ cell counting technology, additional Brightfield analysis features, and fluorescent green and red detection channels.
The SpectraMax® MiniMax™ 300 Imaging Cytometer simplifies workflow by enabling cell and whole-well assays in a small footprint. It eliminates cell staining for cell counting and confluency measurements by using StainFree™ Technology. Multi-parametric cell-based assays with brightfield and green and red fluorescent imaging result in more data. Set up, image, and analyze cells in a snap using SoftMax® Pro Microplate Data Acquisition and Analysis Software. Higher sensitivity permits easier measurement of low cell concentration samples. With lower variance, users can normalize plate data to the number of cells in each well.
The SpectraMax® MiniMax™ 300 Imaging Cytometer supports a multitude of absorbance assay categories including Cell Counting, Cell Viability, Proliferation, and Cytotoxicity; ELISAs and Immunoassays; Enzyme Assays; Endotoxin Detection; Protein Quantitation; Marker Expression; Reporter Gene Assays; Phosphotases/Kinases.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty