Would you like to log in again?
Seldin Periosteal Elevator, OR Grade, Sklar®
Supplier: Sklar
97-0527
10192-264EA 160.41 USD InStock
10192-264
Seldin Periosteal Elevator, OR Grade, Sklar®
Retractors
Sklar's® Seldin Periosteal Elevator is used to remove soft tissue and periosteum, most commonly in oral and plastic surgery.
  • Double-ended and features tips of two different lengths
In oral surgery it can further be used to loosen and remove the tooth.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty