Would you like to log in again?
Phaneuf Uterine Artery Forceps, OR Grade, Sklar
Supplier: Sklar
90-2682 90-2680
10199-138EA 350.06 USD InStock
10199-138 10192-272
Phaneuf Uterine Artery Forceps, OR Grade, Sklar
Forceps
Phaneuf Uterine Artery Forceps are ratcheted, finger ring OB/GYN forceps used for clamping vessels during hysterectomy procedures.
Horizontal serrations and 1 x 2 teeth at the tip provide a secure grasp on tissue. The shanks are straight, but jaws may be straight or curved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty