Would you like to log in again?
Umbilical Scissors, OR Grade, Sklar
Supplier: Sklar
92-5540
10192-312EA 185.31 USD InStock
10192-312
Umbilical Scissors, OR Grade, Sklar
Scissors
Umbilical Scissors are non-ratcheted, finger ring scissors used for cutting the umbilical cord.
These scissors feature wide, flat, curved blades. They are used without clamping the umbilical cord because the blades hold the cord in place. The umbilical cord is often clamped at two points and cut in the middle with standard scissors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty