Would you like to log in again?
Stevens Tenotomy Scissors, OR Grade, Sklar
Supplier: Sklar
64-1341 64-1241 64-1243 64-1342 64-1242 22-1240 64-1343
10192-342EA 144.75 USD InStock
10192-342 10197-850 10192-360 10194-350 10194-352 89025-230 10197-856
Stevens Tenotomy Scissors, OR Grade, Sklar
Scissors
Stevens Tenotomy Scissors are used for delicate dissection and cutting in ophthalmologic, neurological, and plastic surgery procedures.
They have long handles and small, sharp blades with sharp or blunt wedge-shaped tips
Censitrac Ready OR Grade Stevens Tenotomy Scissors are industry standard and constructed of stainless steel.
The long handles allow the surgeon to have a firm grip and control while dissecting or cutting in small or narrow areas. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used more often for dissecting and cutting mucus membranes.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty