Would you like to log in again?
Sklarlite™ Kelly Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sklarlite™
23-2107 23-2108
10192-472EA 133.37 USD InStock
10192-472 10192-348
Sklarlite™ Kelly Hemostatic Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Kelly Hemostatic Forceps are used for clamping large blood vessels or manipulating heavy tissue
They are available curved or straight with a ratcheted finger ring handle. Similar to Rankin forceps, the jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Kelly forceps are shorter and heavier than Rankin forceps. Sklarlite™ instruments are a lighter version of standard Sklar instruments. This lightweight modification provides the surgeon with greater control and less fatigue.
They may also be used for soft tissue dissection.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty