Would you like to log in again?
Vasectomy Piercing Forceps, OR Grade, Sklar
Supplier: Sklar
85-6224
10192-390EA 282.2 USD InStock
10192-390
Vasectomy Piercing Forceps, OR Grade, Sklar
Forceps
Vasectomy Piercing Forceps are a ratcheted, finger ring instrument with sharp tips used to pierce the scrotal skin in sutureless vasectomy procedures
It is used in conjunction with the vasectomy clamp with ring tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty