Would you like to log in again?
Randall Endometrial Biopsy Curette, OR Grade, Sklar
Supplier: Sklar
90-6295
10192-452EA 126.97 USD InStock
10192-452
Randall Endometrial Biopsy Curette, OR Grade, Sklar
Curettes
Sklar's Randall Endometrial Biopsy Curette is an OB/GYN instrument typically used for obtaining tissue samples from the endometrium for biopsy
It is a hollow curette with a thin stylet, and a small, sharp opening near the slightly curved tip. The curette, with the stylet inside, is inserted and moved in such a motion so as to collect tissue from the endometrial cavity. The tissue is then removed from the curette by pulling the stylet out and placing the tissue in a separate container until biopsy. This product has a 4mm outer diameter. There is a small, smooth, oval thumb piece attached to the curette to improve control of the instrument.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty