Would you like to log in again?
Buck Neurohammer, OR Grade, Sklar
Supplier: Sklar
06-3370
10192-460EA 64.01 USD InStock
10192-460
Buck Neurohammer, OR Grade, Sklar
Mallets and Hammers
Buck Neurohammer is a diagnostic tool that features a head design similar to that of the Dejerine Hammer, however the non-latex bumpers on each end of the head are different sizes.
  • Used for reflex and general neurological testing
  • Reusable, latex-free
The Buck Hammer comes with a pin that screws into the top of the handle which protrudes slightly vertically through the middle point of the bumpers. This hammer also contains a brush that screws into the bottom of the handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty