Would you like to log in again?
Coller Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
17-3375 17-3275 17-3365 17-3265 17-3555 17-3455
10194-230EA 125.13 USD InStock
10194-230 10194-790 10194-794 10194-796 10192-466 10194-446
Coller Hemostatic Forceps, OR Grade, Sklar
Forceps
Coller Hemostatic Forceps are ratcheted, finger ring forceps used for grasping tissue and controlling blood flow in vessels during surgical procedures
Censitrac Ready products are industry standard and constructed of stainless steel.
They may be known as Coller-Crile forceps.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty