Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Merit™ Crile Forceps, Physician Grade, Sklar
Supplier: Sklar
97-458 97-463 97-452 97-447
10184-808EA 29.73 USD InStock
10184-808 10193-696 10192-468 10184-806
Merit™ Crile Forceps, Physician Grade, Sklar
Forceps
Merit™ Crile Hemostatic Forceps, also known as a Crile Hemostat, are used for clamping blood vessels or tissue before cauterization or ligation.
Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. They may also be used for soft tissue dissection typically no deeper than 6-8" or in laparotomy procedures.
Merit™ instruments are office-grade line of products and used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty