Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SklarLite™ Half Size Rigid Sterilization Containers, Sklar
Supplier: Sklar
SklarLite™
10-4766 10-4772 10-4702 10-4502 10-4710 10-4704 10-4512 10-4706 10-4658 10-4712 10-4762 10-4516 10-4746 10-4768 10-4750 10-4690 10-4528 10-4694 10-4526 10-4778 10-4532 10-4754 10-4738 10-4534 10-4760 10-4524 10-4742 10-4740 10-4692 10-4774 10-4756 10-4776 10-4744 10-4708 10-4696 10-4504 10-4500 10-4758 10-4770 10-4698 10-4506 10-4782 10-4752 10-4780 10-4700 10-4664 10-4764 10-4662 10-4510 10-4660 10-4536 10-4508 10-4748 10-4520 10-4530 10-4518 10-4514
10192-928EA 1069.78 USD InStock
10192-928 10192-968 10192-924 10192-926 10192-964 10192-920 10192-966 10192-922 10192-960 10192-684 10192-962 10192-686 10192-680 10192-682 10192-916 10192-918 10192-956 10192-912 10192-958 10192-914 10192-678 10192-952 10192-954 10192-910 10192-674 10192-676 10192-950 10192-670 10192-672 10192-906 10192-908 10192-946 10192-948 10192-904 10192-986 10192-942 10192-666 10192-944 10192-668 10192-982 10192-662 10192-940 10192-984 10192-664 10192-980 10192-660 10192-938 10192-934 10192-978 10192-936 10192-930 10192-974 10192-976 10192-932 10192-970 10192-972 10163-488
SklarLite™ Half Size Rigid Sterilization Containers, Sklar
Boxes Transport Boxes
The SklarLite™ Rigid Sterilization Container provides an organized, routine, and secure approach to the sterilization of surgical instruments
  • Sterilization container lid is available in a variety of colours
  • Lids are made of anodized aluminium alloy, these reusable lids are nonsterile and latex-free
Customize the SklarLite™ containers by selecting colored lids and ID labels. Available in a choice of six vibrant colors for easy indentification of instrument sets by procedure and/or department. Various configurations are available to fit surgery center needs.
Accessories information: SklarLite™ Identification Labels are ideally suited for use with the SklarLite™ Rigid Sterilization Containers and Container Systems. Polyester film labels are nonsterile and latex-free. Disposable identification labels are color coded to enable quick and simple sorting and selection.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty