Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sponge/Utility Bowls, Sklar
Supplier: Sklar
10-1502 10-1502-12
10192-802EA 25.03 USD InStock
10192-802 10192-804
Sponge/Utility Bowls, Sklar
Trays/Dishes/Pots Laboratory Trays/Dishes
Sklar Sponge Bowls are most commonly utilized as a recepticle for contaminated sponges to be placed throughout a surgical procedure.
They may; however, also be used as a container for holding sponges or sponge sticks that have been soaked in a sterile solution to be used during a procedure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty