Would you like to log in again?
SklarLite™ Mid Size Rigid Sterilization Container Lids, Sklar
Supplier: Sklar
Sklarlite™
10-4464 10-4476 10-4462 10-4472 10-4454 10-4458 10-4466 10-4470 10-4474 10-4460 10-4456 10-4468
10192-818EA 679.54 USD InStock
10192-818 10192-890 10192-892 10192-880 10192-876 10192-886 10192-878 10192-888 10192-894 10192-882 10192-896 10192-884
SklarLite™ Mid Size Rigid Sterilization Container Lids, Sklar
Lids Lids for Laboratory Vessels/Containers
The SklarLite™ Mid Size Rigid Sterilization Container Lids are available in a variety of colors.
  • Available in perforated or solid formats
Made of anodized aluminum alloy, these reusable lids are nonsterile and latex-free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty