The SklarLite™ Rigid Sterilization Container provides an organized, routine, and secure approach to the sterilization of surgical instruments
- Sterilization container lid is available in a variety of colours
- Lids are made of anodized aluminium alloy, these reusable lids are nonsterile and latex-free
Customize the SklarLite™ containers by selecting colored lids and ID labels. Available in a choice of six vibrant colors for easy indentification of instrument sets by procedure and/or department. Various configurations are available to fit surgery center needs.
Accessories information: SklarLite™ Identification Labels are ideally suited for use with the SklarLite™ Rigid Sterilization Containers and Container Systems. Polyester film labels are nonsterile and latex-free. Disposable identification labels are color coded to enable quick and simple sorting and selection.